N-Lauroylsarcosine sodium salt, 95%
N-Lauroylsarcosine sodium salt, 95%
N-Lauroylsarcosine sodium salt, 95%
N-Lauroylsarcosine sodium salt, 95%
Thermo Scientific Chemicals
N-Lauroylsarcosine sodium salt, 95%
CAS: 137-16-6 | C15H28NNaO3 | 293.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
1 kg
Packaging:
View all
Glass bottle
Catalog number FSA434371000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialN-Lauroylsarcosine sodium salt
CAS137-16-6
CAS Min %94
Health Hazard 1Serious eye damage/eye irritation (category 1), Acute toxicity (category 2), Skin corrosion/irritation (category 2)
CAS Max %100