Morpholine, 99.5%, purified by redistillation, AcroSeal™
Morpholine, 99.5%, purified by redistillation, AcroSeal™
Morpholine, 99.5%, purified by redistillation, AcroSeal™
Morpholine, 99.5%, purified by redistillation, AcroSeal™
Thermo Scientific Chemicals
Morpholine, 99.5%, purified by redistillation, AcroSeal™
CAS: 110-91-8 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number FSA433851000
Price (MYR)
400.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
400.00
EA
Add to cart
Specifications
Chemical Name or MaterialMorpholine
CAS110-91-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if inhaled.
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (