1-Chlorobutane, 99.5%, anhydrous, AcroSeal™
1-Chlorobutane, 99.5%, anhydrous, AcroSeal™
1-Chlorobutane, 99.5%, anhydrous, AcroSeal™
1-Chlorobutane, 99.5%, anhydrous, AcroSeal™
Thermo Scientific Chemicals
1-Chlorobutane, 99.5%, anhydrous, AcroSeal™
CAS: 109-69-3 | C4H9Cl | 92.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA433820010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Chlorobutane
CAS109-69-3
CAS Min %99.4
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0