Ethylbenzene, 99.8%, anhydrous, AcroSeal™
Ethylbenzene, 99.8%, anhydrous, AcroSeal™
Ethylbenzene, 99.8%, anhydrous, AcroSeal™
Ethylbenzene, 99.8%, anhydrous, AcroSeal™
Ethylbenzene, 99.8%, anhydrous, AcroSeal™
Thermo Scientific Chemicals
Ethylbenzene, 99.8%, anhydrous, AcroSeal™
CAS: 100-41-4 | C8H10 | 106.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA433801000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEthylbenzene
CAS100-41-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful to aquatic life with long lasti
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a