Methylcyclohexane, 99%, for spectroscopy
Methylcyclohexane, 99%, for spectroscopy
Methylcyclohexane, 99%, for spectroscopy
Methylcyclohexane, 99%, for spectroscopy
Thermo Scientific Chemicals
Methylcyclohexane, 99%, for spectroscopy
CAS: 108-87-2 | C7H14 | 98.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA433770010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethylcyclohexane
CAS108-87-2
Health Hazard 1GHS Signal Word: Danger
Melting Point-126.3°C
Density0.7700g/mL
View more
For UV/VIS spectroscopy
RUO – Research Use Only