1,2-Dichloroethane, 99.8%, for HPLC
1,2-Dichloroethane, 99.8%, for HPLC
1,2-Dichloroethane, 99.8%, for HPLC
1,2-Dichloroethane, 99.8%, for HPLC
1,2-Dichloroethane, 99.8%, for HPLC
Thermo Scientific Chemicals
1,2-Dichloroethane, 99.8%, for HPLC
CAS: 107-06-2 | C2H4Cl2 | 98.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA433580010
Price (MYR)
749.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
749.00
EA
Add to cart
Specifications
Chemical Name or Material1,2-Dichloroethane
Melting Point-35.0°C
Absorbance0.005 max. at 260nm, 0.005 max. at 270nm, 0.01 max. at 300nm, 0.01 max. at 400nm, 0.05 max. at 250nm, 0.15 max. at 240nm, 1 max. at 230nm
CAS107-06-2
ColorWhite
View more
For HPLC
RUO – Research Use Only