Check Order Status
5-Benzyloxyindole-3-carboxaldehyde, 98%, Thermo Scientific™
Element Search
Structure Search
5-Benzyloxyindole-3-carboxaldehyde, 98%, Thermo Scientific™
Have Questions?
Catalog Number
Quantity
Packaging
FSA433430010
1 g
Glass bottle
Catalog number
FSA433430010
Price (MYR)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material
5-Benzyloxyindole-3-carboxaldehyde
CAS
6953-22-6
CAS Min %
97.5
Health Hazard 1
GHS Signal Word:
Warning
CAS Max %
100.0