Check Order Status
2-(Trifluoromethoxy)benzaldehyde, 96%, Thermo Scientific™
Element Search
Structure Search
2-(Trifluoromethoxy)benzaldehyde, 96%, Thermo Scientific™
Have Questions?
Change view
Quantity:
5 g
1 g
Catalog number
FSA433200050
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material
2-(Trifluoromethoxy)benzaldehyde
CAS
94651-33-9
CAS Min %
95.0
Health Hazard 1
GHS Signal Word:
Warning
CAS Max %
100.0