Tetrazole, 3 to 4 wt.% solution in acetonitrile, AcroSeal™
Tetrazole, 3 to 4 wt.% solution in acetonitrile, AcroSeal™
Tetrazole, 3 to 4 wt.% solution in acetonitrile, AcroSeal™
Tetrazole, 3 to 4 wt.% solution in acetonitrile, AcroSeal™
Tetrazole, 3 to 4 wt.% solution in acetonitrile, AcroSeal™
Thermo Scientific Chemicals
Tetrazole, 3 to 4 wt.% solution in acetonitrile, AcroSeal™
CAS: 288-94-8 | CH2N4 | 70.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA432991000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS288-94-8, 75-05-8
Specifications
Chemical Name or MaterialTetrazole
CAS288-94-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapour.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
CAS75-05-8