4-Methoxybenzenesulfonamide, 95%
4-Methoxybenzenesulfonamide, 95%
4-Methoxybenzenesulfonamide, 95%
4-Methoxybenzenesulfonamide, 95%
Thermo Scientific Chemicals
4-Methoxybenzenesulfonamide, 95%
CAS: 1129-26-6 | C7H9NO3S | 187.213 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA432910250
Price (MYR)
1,699.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,699.00
EA
Add to cart
Specifications
Chemical Name or Material4-Methoxybenzenesulfonamide
CAS1129-26-6
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0