Methyltrimethoxysilane, 97%, AcroSeal™, Thermo Scientific Chemicals
Methyltrimethoxysilane, 97%, AcroSeal™, Thermo Scientific Chemicals
Methyltrimethoxysilane, 97%, AcroSeal™, Thermo Scientific Chemicals
Methyltrimethoxysilane, 97%, AcroSeal™, Thermo Scientific Chemicals
Methyltrimethoxysilane, 97%, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyltrimethoxysilane, 97%, AcroSeal™, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA432641000
Price (MYR)
205.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
205.00
EA
Add to cart
Specifications
Chemical Name or MaterialMethyltrimethoxysilane
CAS1185-55-3
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0