2-Amino-4-phenylthiazole, 97%
2-Amino-4-phenylthiazole, 97%
2-Amino-4-phenylthiazole, 97%
2-Amino-4-phenylthiazole, 97%
Thermo Scientific Chemicals
2-Amino-4-phenylthiazole, 97%
CAS: 2010-06-2 | C9H8N2S | 176.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA432490050
Price (MYR)
1,418.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,418.00
EA
Add to cart
Specifications
Chemical Name or Material2-Amino-4-phenylthiazole
CAS2010-06-2
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0