6-Iodo-1H-indazole, 97%, Thermo Scientific™
6-Iodo-1H-indazole, 97%, Thermo Scientific™
6-Iodo-1H-indazole, 97%, Thermo Scientific™
6-Iodo-1H-indazole, 97%, Thermo Scientific™
Thermo Scientific Chemicals
6-Iodo-1H-indazole, 97%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA432420010
Price (MYR)
1,421.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,421.00
EA
Add to cart
Specifications
Chemical Name or Material6-Iodo-1H-indazole
CAS261953-36-0
CAS Min %96.0
CAS Max %100.0
Health Hazard 2GHS H Statement: