Trifluoroacetic acid, 99+%, for HPLC
Trifluoroacetic acid, 99+%, for HPLC
Trifluoroacetic acid, 99+%, for HPLC
Trifluoroacetic acid, 99+%, for HPLC
Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, for HPLC
CAS: 76-05-1 | C2HF3O2 | 114.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
View all
Glass Bottle
Catalog number FSA432295000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Absorbance0.03 max. at 300 nm, 0.03 max. at 320 nm, 0.04 max. at 290 nm, 0.05 max. at 280 nm, 0.1 max. at 270nm, 1 max. at 260 nm
Chemical Name or MaterialTrifluoroacetic Acid
Name NoteFor HPLC
CAS76-05-1
Melting Point-15.0°C
View more
For HPLC
RUO – Research Use Only