Decyldimethylchlorosilane, 97%, Thermo Scientific™
Decyldimethylchlorosilane, 97%, Thermo Scientific™
Decyldimethylchlorosilane, 97%, Thermo Scientific™
Decyldimethylchlorosilane, 97%, Thermo Scientific™
Thermo Scientific Chemicals
Decyldimethylchlorosilane, 97%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number FSA432190250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDecyldimethylchlorosilane
CAS38051-57-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat