Tetrabutylammonium chloride hydrate, 96%
Tetrabutylammonium chloride hydrate, 96%
Tetrabutylammonium chloride hydrate, 96%
Tetrabutylammonium chloride hydrate, 96%
Thermo Scientific Chemicals
Tetrabutylammonium chloride hydrate, 96%
Phase transfer catalyst | CAS: 37451-68-6 | C16H36ClN | 277.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass Bottle
Catalog number FSA432061000
Price (MYR)
1,959.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,959.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetrabutylammonium Chloride Hydrate
CAS37451-68-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
View more
Phase transfer catalyst
RUO – Research Use Only