Oxolinic acid, 98%
Oxolinic acid, 98%
Oxolinic acid, 98%
Thermo Scientific Chemicals
Oxolinic acid, 98%
CAS: 14698-29-4 | C13H11NO5 | 261.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA431510050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialOxolinic acid
CAS14698-29-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Harmful if swallowed.
Health Hazard 3GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.