3-Aminopropyltriethoxysilane, 99%, AcroSeal™
3-Aminopropyltriethoxysilane, 99%, AcroSeal™
3-Aminopropyltriethoxysilane, 99%, AcroSeal™
3-Aminopropyltriethoxysilane, 99%, AcroSeal™
Thermo Scientific Chemicals
3-Aminopropyltriethoxysilane, 99%, AcroSeal™
CAS: 919-30-2 | C9H23NO3Si | 221.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA430948000
Price (MYR)
2,212.00
EA
Add to cart
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
2,212.00
EA
Add to cart
Specifications
Chemical Name or Material3-Aminopropyltriethoxysilane
CAS919-30-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0