1,1,1,3,3,3-Hexamethyldisilazane, 98%, AcroSeal™
1,1,1,3,3,3-Hexamethyldisilazane, 98%, AcroSeal™
1,1,1,3,3,3-Hexamethyldisilazane, 98%, AcroSeal™
1,1,1,3,3,3-Hexamethyldisilazane, 98%, AcroSeal™
1,1,1,3,3,3-Hexamethyldisilazane, 98%, AcroSeal™
Thermo Scientific Chemicals
1,1,1,3,3,3-Hexamethyldisilazane, 98%, AcroSeal™
CAS: 999-97-3 | C6H19NSi2 | 161.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA430851000
Price (MYR)
300.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
300.00
BOT
Add to cart
Specifications
Chemical Name or Material1,1,1,3,3,3-Hexamethyldisilazane
CAS999-97-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
Silylation reagent
RUO – Research Use Only