2-Amino-6-chloropyridine, 97%
2-Amino-6-chloropyridine, 97%
2-Amino-6-chloropyridine, 97%
2-Amino-6-chloropyridine, 97%
Thermo Scientific Chemicals
2-Amino-6-chloropyridine, 97%
CAS: 45644-21-1 | C5H5ClN2 | 128.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA429500050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Amino-6-chloropyridine
CAS45644-21-1
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0