Amberlite™ IRN-78, OH-form, ion-exchange resin
Amberlite™ IRN-78, OH-form, ion-exchange resin
Amberlite™ IRN-78, OH-form, ion-exchange resin
Amberlite™ IRN-78, OH-form, ion-exchange resin
Thermo Scientific Chemicals
Amberlite™ IRN-78, OH-form, ion-exchange resin
CAS: 11128-95-3
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA429295000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmberlite™ IRN-78 ion-exchange resin
Physical FormSolution
Name NoteIon-exchange Resin, OH-form
CAS11128-95-3
Health Hazard 1Danger
View more
Ion exchange resin
RUO – Research Use Only