Propylene carbonate, 99.5%, anhydrous, AcroSeal™
Propylene carbonate, 99.5%, anhydrous, AcroSeal™
Propylene carbonate, 99.5%, anhydrous, AcroSeal™
Propylene carbonate, 99.5%, anhydrous, AcroSeal™
Propylene carbonate, 99.5%, anhydrous, AcroSeal™
Propylene carbonate, 99.5%, anhydrous, AcroSeal™
Thermo Scientific Chemicals
Propylene carbonate, 99.5%, anhydrous, AcroSeal™
CAS: 108-32-7 | C4H6O3 | 102.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA429261000
Price (MYR)
371.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
371.00
EA
Add to cart
Specifications
Chemical Name or MaterialPropylene carbonate
CAS108-32-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
Health Hazard 3GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
View more
Suitable for battery materials development.
RUO – Research Use Only