Lithium borohydride, 4M (10 wt.%) solution in THF, AcroSeal™
Lithium borohydride, 4M (10 wt.%) solution in THF, AcroSeal™
Lithium borohydride, 4M (10 wt.%) solution in THF, AcroSeal™
Lithium borohydride, 4M (10 wt.%) solution in THF, AcroSeal™
Lithium borohydride, 4M (10 wt.%) solution in THF, AcroSeal™
Thermo Scientific Chemicals
Lithium borohydride, 4M (10 wt.%) solution in THF, AcroSeal™
CAS: 16949-15-8 | BH4Li | 21.78 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA428881000
Price (MYR)
1,208.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
1,208.00
EA
Add to cart
Chemical Identifiers
CAS16949-15-8, 109-99-9
Specifications
Chemical Name or MaterialLithium Borohydride
CAS16949-15-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritati
CAS109-99-9
View more
Reduction reagent
RUO – Research Use Only