Tributyl borate, 98%, AcroSeal™
Tributyl borate, 98%, AcroSeal™
Tributyl borate, 98%, AcroSeal™
Tributyl borate, 98%, AcroSeal™
Tributyl borate, 98%, AcroSeal™
Thermo Scientific Chemicals
Tributyl borate, 98%, AcroSeal™
CAS: 688-74-4 | C12H27BO3 | 230.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA428661000
Price (MYR)
345.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
345.00
EA
Add to cart
Specifications
Chemical Name or MaterialTributyl borate
CAS688-74-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a