Methyl cellulose, viscosity 400 cP (2% solution in water)
Methyl cellulose, viscosity 400 cP (2% solution in water)
Methyl cellulose, viscosity 400 cP (2% solution in water)
Methyl cellulose, viscosity 400 cP (2% solution in water)
Thermo Scientific Chemicals
Methyl cellulose, viscosity 400 cP (2% solution in water)
CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass bottle
Catalog number FSA428432500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl cellulose
Name NoteViscosity 400 cP (2% solution in water)
CAS9004-67-5
PackagingGlass bottle
Molecular FormulaC20H38O11