Ammonia, ca. 7N solution in methanol, AcroSeal™
Ammonia, ca. 7N solution in methanol, AcroSeal™
Ammonia, ca. 7N solution in methanol, AcroSeal™
Ammonia, ca. 7N solution in methanol, AcroSeal™
Ammonia, ca. 7N solution in methanol, AcroSeal™
Thermo Scientific Chemicals
Ammonia, ca. 7N solution in methanol, AcroSeal™
CAS: 7664-41-7 | H3N | 17.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Catalog number FSA428381000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS7664-41-7, 67-56-1
Specifications
Chemical Name or MaterialAmmonia
ColorColorless to Yellow
Name Note7N solution in methanol
CAS7664-41-7
Health Hazard 1GHS Signal Word: Danger