Titanium(IV) chloride, 0.09M solution in 20% HCl, AcroSeal™
Titanium(IV) chloride, 0.09M solution in 20% HCl, AcroSeal™
Titanium(IV) chloride, 0.09M solution in 20% HCl, AcroSeal™
Titanium(IV) chloride, 0.09M solution in 20% HCl, AcroSeal™
Titanium(IV) chloride, 0.09M solution in 20% HCl, AcroSeal™
Thermo Scientific Chemicals
Titanium(IV) chloride, 0.09M solution in 20% HCl, AcroSeal™
CAS: 7550-45-0 | Cl4Ti | 189.71 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA428351000100 mLAcroSeal™ Glass Bottle
Catalog number FSA428351000
Price (MYR)
478.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
478.00
EA
Add to cart
Chemical Identifiers
CAS7550-45-0, 7647-01-0, 7732-18-5
Specifications
Chemical Name or MaterialTitanium(IV) chloride
CAS7550-45-0
ColorColorless to Yellow
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
View more
Lewis acid
RUO – Research Use Only