Di(propylene glycol) methyl ether, 99%, pure, mixture of isomers
Di(propylene glycol) methyl ether, 99%, pure, mixture of isomers
Di(propylene glycol) methyl ether, 99%, pure, mixture of isomers
Di(propylene glycol) methyl ether, 99%, pure, mixture of isomers
Thermo Scientific Chemicals
Di(propylene glycol) methyl ether, 99%, pure, mixture of isomers
CAS: 34590-94-8 | C7H16O3 | 148.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
1 L
Packaging:
View all
Plastic drum
Catalog number FSA428200050
Price (MYR)
1,287.00
EA
Add to cart
Quantity:
5 L
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
1,287.00
EA
Add to cart
Specifications
Chemical Name or MaterialDi(propylene glycol) methyl ether
Name NoteMixture of isomers
CAS34590-94-8
Melting Point-80.0°C
Density0.9500g/mL