Methyl linoleate, 99%
Methyl linoleate, 99%
Methyl linoleate, 99%
Methyl linoleate, 99%
Thermo Scientific Chemicals
Methyl linoleate, 99%
CAS: 112-63-0 | C19H34O2 | 294.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA428130250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl linoleate
CAS112-63-0
Melting Point-35.0°C
Density0.8900g/mL
EINECS Number203-993-