Ethylaluminium dichloride, 1.8M solution in toluene, AcroSeal™
Ethylaluminium dichloride, 1.8M solution in toluene, AcroSeal™
Ethylaluminium dichloride, 1.8M solution in toluene, AcroSeal™
Ethylaluminium dichloride, 1.8M solution in toluene, AcroSeal™
Thermo Scientific Chemicals
Ethylaluminium dichloride, 1.8M solution in toluene, AcroSeal™
CAS: 563-43-9 | C2H5AlCl2 | 126.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number FSA428041000
Price (MYR)
690.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
690.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthylaluminium dichloride
Name Note1.8M solution in toluene
CAS563-43-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
May ca
View more
Lewis acid
RUO – Research Use Only