tert-Butyl methyl ether, 99.8%, for spectroscopy ACS, meets the requirements of Reag.Ph.Eur.
tert-Butyl methyl ether, 99.8%, for spectroscopy ACS, meets the requirements of Reag.Ph.Eur.
tert-Butyl methyl ether, 99.8%, for spectroscopy ACS, meets the requirements of Reag.Ph.Eur.
tert-Butyl methyl ether, 99.8%, for spectroscopy ACS, meets the requirements of Reag.Ph.Eur.
Thermo Scientific Chemicals
tert-Butyl methyl ether, 99.8%, for spectroscopy ACS, meets the requirements of Reag.Ph.Eur.
CAS: 1634-04-4 | C5H12O | 88.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA4280100252.5 LGlass Bottle
Catalog number FSA428010025
Price (MYR)
665.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
665.00
EA
Add to cart
Specifications
Chemical Name or Materialtert-Butyl methyl ether
Melting Point-110.0°C
Name NoteFor spectroscopy
Absorbance1 cm cell vs HPLC-grade water, 0.097 max. at 255 nm, 0.009 max. at 280 nm, 1 cm cell vs HPLC-grade water at 0 nm
CAS1634-04-4
View more
for UV/VIS spectroscopy
RUO – Research Use Only