Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™
Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™
Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™
Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™
Thermo Scientific Chemicals
Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™
CAS: 23519-77-9 | C12H28O4Zr | 327.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA427938000
Price (MYR)
1,368.00
EA
Add to cart
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
1,368.00
EA
Add to cart
Specifications
Chemical Name or MaterialZirconium(IV) propoxide
Name Noteca. 70%
CAS23519-77-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye damage.
Flammable liquid and vapor.