7-Hydroxycoumarinyl-4-acetic acid, 97%
7-Hydroxycoumarinyl-4-acetic acid, 97%
7-Hydroxycoumarinyl-4-acetic acid, 97%
7-Hydroxycoumarinyl-4-acetic acid, 97%
Thermo Scientific Chemicals
7-Hydroxycoumarinyl-4-acetic acid, 97%
CAS: 6950-82-9 | C11H8O5 | 220.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA427900010
Price (MYR)
456.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
456.00
EA
Add to cart
Specifications
Chemical Name or Material7-Hydroxycoumarinyl-4-acetic acid
CAS6950-82-9
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0