cis-4,7,10,13,16,19-Docosahexaenoic acid, 98%
cis-4,7,10,13,16,19-Docosahexaenoic acid, 98%
cis-4,7,10,13,16,19-Docosahexaenoic acid, 98%
cis-4,7,10,13,16,19-Docosahexaenoic acid, 98%
Thermo Scientific Chemicals
cis-4,7,10,13,16,19-Docosahexaenoic acid, 98%
CAS: 6217-54-5 | C22H32O2 | 328.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Packaging:
View all
Glass Bottle
Catalog number FSA427641000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Materialcis-4,7,10,13,16,19-Docosahexaenoic acid
CAS6217-54-5
Melting Point-44°C
ColorColorless to Yellow
Density0.9500g/mL