Isophorone diisocyanate, 98+%, mixture of isomers
Isophorone diisocyanate, 98+%, mixture of isomers
Isophorone diisocyanate, 98+%, mixture of isomers
Isophorone diisocyanate, 98+%, mixture of isomers
Isophorone diisocyanate, 98+%, mixture of isomers
Thermo Scientific Chemicals
Isophorone diisocyanate, 98+%, mixture of isomers
CAS: 4098-71-9 | C12H18N2O2 | 222.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
50 mL
1 L
Packaging:
Glass bottle
Catalog number FSA427602500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialIsophorone diisocyanate
CAS4098-71-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic to aquatic life with long lasti
Health Hazard 3GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If skin irritation or