Tetrahydro-beta-carboline, 98%
Tetrahydro-beta-carboline, 98%
Tetrahydro-beta-carboline, 98%
Tetrahydro-beta-carboline, 98%
Thermo Scientific Chemicals
Tetrahydro-beta-carboline, 98%
CAS: 16502-01-5 | C11H13ClN2 | 208.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA427580050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTetrahydro-β-carboline
CAS16502-01-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi