N-Acetyl-D-mannosamine, 98%, Thermo Scientific Chemicals
N-Acetyl-D-mannosamine, 98%, Thermo Scientific Chemicals
N-Acetyl-D-mannosamine, 98%, Thermo Scientific Chemicals
N-Acetyl-D-mannosamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Acetyl-D-mannosamine, 98%, Thermo Scientific Chemicals
N-Acetyl-D-mannosamine (ManNAc), CAS # 7772-94-3, is a monosaccharide sialic acid precursor.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA427530050
Price (MYR)
1,611.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,611.00
EA
Add to cart
Specifications
Chemical Name or MaterialN-Acetyl-D-mannosamine
CAS7772-94-3
CAS Min %97.5
CAS Max %100.0
Health Hazard 2GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
General Description
  • N-Acetyl-D-mannosamine (ManNAc) is a precursor used to synthesize N-acetylneuraminic acid, the most common physiologic sialic acid present in humans and other mammals
  • Sialic acids are terminal sugars on glycoconjugates that can modify biochemistry of glycoproteins and lipids, mediate cellular adhesion, and act as immune regulators
  • Defects in sialic acid biosynthesis are associated with a number of pathologies
  • Sialic acids are often found at increased expression levels on tumor cell surfaces
Applications
  • As a sialic acid precursor, N-Acetyl-D-mannosamine can be used in many glycoengineering applications, such as modifying immune cell activity, imaging glycans, or blocking pathogen binding
  • N-Acetyl-D-mannosamine hasbeen used in preclinical laboratory studies as a treatment for hereditary inclusion body myopathy, a progressive neuromuscular disease via increased sialylation of podocalyxin
  • N-Acetyl-D-mannosamine has been used in laboratory studies to induce IgG sialylation and prevent obesity-induced hypertension in mice following high-fat diet
RUO – Research Use Only
General References:
  1. Peng, J.; Vongpatanasin, W.; Sacharidou, A.; Kifer, D.; Yuhanna, I.S.; Banerjee, S.; Tanigaki, K.; Polasek, O.; Chu, H.; Sundgren, N.C.; Rohatgi, A.; Chambliss, K.L.; Lauc, G.; Mineo, C.; Shaul, P.W. Supplementation With the Sialic Acid Precursor N-Acetyl-D-Mannosamine Breaks the Link Between Obesity and Hypertension. Circulation. 2019, 140(24), 2005-2018.
  2. Moons, S.J.; Adema, G.J.; Derks, M.T.; Boltje, T.J.; Büll, C. Sialic acid glycoengineering using N-acetylmannosamine and sialic acid analogs. Glycobiology. 2019, 29(6), 433-445.
  3. Büll, C.; Stoel, M.A.; den Brok, M.H.; Adema, GJ. Sialic acids sweeten a tumor's life. Cancer Res. 2014, 74(12), 3199-204.
  4. Galeano, B.; Klootwijk, R.; Manoli, I.; Sun, M.; Ciccone, C.; Darvish, D.; Starost, M.F.; Zerfas, P.M.; Hoffmann, V.J.; Hoogstraten-Miller, S.; Krasnewich, D.M.; Gahl, W.A.; Huizing, M. Mutation in the key enzyme of sialic acid biosynthesis causes severe glomerular proteinuria and is rescued by N-acetylmannosamine. J Clin Invest. 2007, 117(6), 1585-94.