tert-Butylamine, 99%, AcroSeal™
tert-Butylamine, 99%, AcroSeal™
tert-Butylamine, 99%, AcroSeal™
tert-Butylamine, 99%, AcroSeal™
tert-Butylamine, 99%, AcroSeal™
Thermo Scientific Chemicals
tert-Butylamine, 99%, AcroSeal™
CAS: 75-64-9 | C4H11N | 73.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number FSA427241000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialtert-Butylamine
CAS75-64-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Health Hazard 3GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye p