Boron trifluoride etherate, ca. 48% BF3, AcroSeal™
Boron trifluoride etherate, ca. 48% BF3, AcroSeal™
Boron trifluoride etherate, ca. 48% BF3, AcroSeal™
Boron trifluoride etherate, ca. 48% BF3, AcroSeal™
Thermo Scientific Chemicals
Boron trifluoride etherate, ca. 48% BF3, AcroSeal™
CAS: 109-63-7 | C4H10BF3O | 141.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA427118000
Price (MYR)
825.00
EA
Add to cart
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
825.00
EA
Add to cart
Specifications
Chemical Name or MaterialBoron trifluoride etherate
Name Noteca. 48% BF3
CAS109-63-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Causes damage to organs through prolonged or repeated exposure if inhaled.
View more
Lewis acid
RUO – Research Use Only