Chloroform-d, for NMR, 0.02-0.04 v/v% TMS, 99.8 atom % D, AcroSeal™
Chloroform-d, for NMR, 0.02-0.04 v/v% TMS, 99.8 atom % D, AcroSeal™
Chloroform-d, for NMR, 0.02-0.04 v/v% TMS, 99.8 atom % D, AcroSeal™
Chloroform-d, for NMR, 0.02-0.04 v/v% TMS, 99.8 atom % D, AcroSeal™
Chloroform-d, for NMR, 0.02-0.04 v/v% TMS, 99.8 atom % D, AcroSeal™
Thermo Scientific Chemicals
Chloroform-d, for NMR, 0.02-0.04 v/v% TMS, 99.8 atom % D, AcroSeal™
CAS: 865-49-6 | CHCl3 | 120.375 g/mol
Have Questions?
Catalog NumberQuantity
FSA426961000100 mL
Catalog number FSA426961000
Price (MYR)
439.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
439.00
EA
Add to cart
Specifications
Chemical Name or MaterialChloroform-d
Density1.5000g/mL
Deuterium (D)99.75% min.
Physical FormLiquid
Water0.02% max.
View more
For NMR
RUO – Research Use Only