Methyl sulfoxide-d6, for NMR, 99.9 atom% D, AcroSeal™
Methyl sulfoxide-d6, for NMR, 99.9 atom% D, AcroSeal™
Methyl sulfoxide-d6, for NMR, 99.9 atom% D, AcroSeal™
Methyl sulfoxide-d6, for NMR, 99.9 atom% D, AcroSeal™
Methyl sulfoxide-d6, for NMR, 99.9 atom% D, AcroSeal™
Thermo Scientific Chemicals
Methyl sulfoxide-d6, for NMR, 99.9 atom% D, AcroSeal™
CAS: 2206-27-1 | C2H6OS | 84.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA426941000100 mLAcroSeal™ Glass Bottle
Catalog number FSA426941000
Price (MYR)
2,462.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
2,462.00
EA
Add to cart
Specifications
Chemical Name or MaterialMethyl sulfoxide-d6
Name NoteFor NMR, 99.9 atom% D
CAS2206-27-1
Melting Point20.2°C
ColorColorless
View more
For NMR
RUO – Research Use Only