Di-tert-butyl dicarbonate, 1M solution in THF, AcroSeal™
Di-tert-butyl dicarbonate, 1M solution in THF, AcroSeal™
Di-tert-butyl dicarbonate, 1M solution in THF, AcroSeal™
Di-tert-butyl dicarbonate, 1M solution in THF, AcroSeal™
Thermo Scientific Chemicals
Di-tert-butyl dicarbonate, 1M solution in THF, AcroSeal™
CAS: 24424-99-5 | C10H18O5 | 218.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA426801000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS24424-99-5, 109-99-9
Specifications
Chemical Name or MaterialDi-tert-butyl dicarbonate
Name Note1M Solution in THF
CAS24424-99-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Fatal if inhaled.
May cause drowsiness