4-tert-Butylbenzaldehyde, 95%
4-tert-Butylbenzaldehyde, 95%
4-tert-Butylbenzaldehyde, 95%
4-tert-Butylbenzaldehyde, 95%
Thermo Scientific Chemicals
4-tert-Butylbenzaldehyde, 95%
CAS: 939-97-9 | C11H14O | 162.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
Packaging:
Glass bottle
Catalog number FSA426520500
Price (MYR)
531.00
EA
Add to cart
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
531.00
EA
Add to cart
Specifications
Chemical Name or Material4-tert-Butylbenzaldehyde
CAS939-97-9
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0