1,4-Pentadien-3-ol, 97%, stabilized
1,4-Pentadien-3-ol, 97%, stabilized
1,4-Pentadien-3-ol, 97%, stabilized
1,4-Pentadien-3-ol, 97%, stabilized
Thermo Scientific Chemicals
1,4-Pentadien-3-ol, 97%, stabilized
CAS: 922-65-6 | C5H8O | 84.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA426240050
Price (MYR)
677.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
677.00
EA
Add to cart
Specifications
Chemical Name or Material1, 4-Pentadien-3-ol
Name NoteStabilized
CAS922-65-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning