Ammonium sulfide, 20% solution in water
Ammonium sulfide, 20% solution in water
Ammonium sulfide, 20% solution in water
Ammonium sulfide, 20% solution in water
Thermo Scientific Chemicals
Ammonium sulfide, 20% solution in water
CAS: 12135-76-1 | H8N2S | 68.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
Glass bottle
Catalog number FSA426081000
Price (MYR)
1,054.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,054.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium sulfide
Name Note20% solution in water
CAS12135-76-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Very toxic to aquatic life.
Contact with acids liberates toxic gas.