1-Propynylmagnesium bromide, 0.5M solution in THF, AcroSeal™
1-Propynylmagnesium bromide, 0.5M solution in THF, AcroSeal™
1-Propynylmagnesium bromide, 0.5M solution in THF, AcroSeal™
1-Propynylmagnesium bromide, 0.5M solution in THF, AcroSeal™
1-Propynylmagnesium bromide, 0.5M solution in THF, AcroSeal™
Thermo Scientific Chemicals
1-Propynylmagnesium bromide, 0.5M solution in THF, AcroSeal™
CAS: 16466-97-0 | C3H3BrMg | 143.27 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA426071000100 mLAcroSeal™ Glass bottle
Catalog number FSA426071000
Price (MYR)
710.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
710.00
EA
Add to cart
Specifications
Chemical Name or Material1-Propynylmagnesium bromide
CAS16466-97-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
May form explosive peroxides.
Reacts violentl
CAS109-99-9