6-Carboxyfluorescein, 96%
6-Carboxyfluorescein, 96%
6-Carboxyfluorescein, 96%
6-Carboxyfluorescein, 96%
Thermo Scientific Chemicals
6-Carboxyfluorescein, 96%
CAS: 3301-79-9 | C21H12O7 | 376.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
100 mg
Packaging:
Glass bottle
Catalog number FSA425805000
Price (MYR)
4,542.00
EA
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,542.00
EA
Add to cart
Specifications
Chemical Name or Material6-Carboxyfluorescein
CAS3301-79-9
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0