3,5-Dihydroxybenzaldehyde, 98%
3,5-Dihydroxybenzaldehyde, 98%
3,5-Dihydroxybenzaldehyde, 98%
3,5-Dihydroxybenzaldehyde, 98%
Thermo Scientific Chemicals
3,5-Dihydroxybenzaldehyde, 98%
CAS: 26153-38-8 | C7H6O3 | 138.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA425650050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3, 5-Dihydroxybenzaldehyde
CAS26153-38-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0