2-(4-Pyridyl)malondialdehyde, 95%
2-(4-Pyridyl)malondialdehyde, 95%
2-(4-Pyridyl)malondialdehyde, 95%
2-(4-Pyridyl)malondialdehyde, 95%
Thermo Scientific Chemicals
2-(4-Pyridyl)malondialdehyde, 95%
CAS: 51076-46-1 | C8H7NO2 | 149.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA4253400252.5 gGlass bottle
Catalog number FSA425340025
Price (MYR)
1,056.00
BOT
Add to cart
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,056.00
BOT
Add to cart
Specifications
Chemical Name or Material2-(4-Pyridyl)malondialdehyde
Name Note95%
CAS51076-46-1
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning