View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL+-Tartaric acid, disodium salt, dihydrate
pH7.0 to 9.0 (5% soln. at 25°C)
CAS6106-24-7
ColorWhite
Assay Percent Range99+%